Learn More
R&D Systems™ Recombinant Human ADAM15 Ecto Domain Western Blot Standard
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ WBC027
117.49 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18158480
  • 150.00€ / Each
Estimated Shipment: 17-12-2024
Description
The Recombinant Human ADAM15 Ecto Domain Western Blot Standard has been validated for the following applications: Western Blot.
Specifications
Specifications
8751
1 Vial
Unconjugated
ADAM15 Western Blot Standard
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Recombinant
Product Suggestions