missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Alpha-mannosidase II GST (N-Term) Protein

Recombinant Protein

Brand:  Novus Biologicals™ H00004124-Q01-25ug

 View more versions of this product

Product Code. 18302128

  • 801.00€ / 25µg
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications

Specifications

Western Blot, ELISA, Protein Array, Immunoaffinity Purification
4124
25 μg
alpha-mannosidase 2, AMan II, EC 3.2.1, EC 3.2.1.114, Golgi alpha-mannosidase II, golgi integral membrane protein 7, GOLIM7, Man II, MANA2MANII, Mannosidase alpha class 2A member 1, mannosidase, alpha type II, mannosidase, alpha, class 2A, member 1, Mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase
>80% by SDS-PAGE and Coomassie blue staining
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
36.74 kDa
Store at -80°C. Avoid freeze-thaw cycles.
MAN2A1
Alpha-mannosidase II
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.