Learn More
Novus Biologicals™ Recombinant Human ATPase Na+/K+ beta 2 His Protein
Recombinant Protein
196.00€ - 2200.00€
Specifications
Concentration | 0.5 mg/ml |
---|---|
For Use With (Application) | Bioactivity, SDS-PAGE |
Formulation | PBS (pH 7.4), 10% glycerol |
Gene ID (Entrez) | 482 |
Molecular Weight (g/mol) | M.W. Theoretical: 26.4 kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18366976
|
Novus Biologicals™
NBP3-15116-50UG |
50 ÎĽg |
470.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18358455
|
Novus Biologicals™
NBP3-15116-500UG |
500 ÎĽg |
2200.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18310917
|
Novus Biologicals™
NBP3-15116-250UG |
250 ÎĽg |
1305.00€
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18399804
|
Novus Biologicals™
NBP3-15116-20UG |
20 ÎĽg |
351.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18304633
|
Novus Biologicals™
NBP3-15116-10UG |
10 ÎĽg |
196.00€
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18383782
|
Novus Biologicals™
NBP3-15116-100UG |
100 ÎĽg |
768.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specific activity is > 3,000 pmol/min/ug, and is defined as the amount of enzyme that hydrolyze 1.0 pmole of Adenosine 5-triphosphate to phosphate per minute per minute at pH 7.5 at 25C.Specifications
0.5 mg/ml | |
PBS (pH 7.4), 10% glycerol | |
M.W. Theoretical: 26.4 kDa | |
<1 EU per 1ug of protein (determined by LAL method) | |
ATP1B2 | |
>90%, by SDS-PAGE |
Bioactivity, SDS-PAGE | |
482 | |
Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles. | |
adhesion molecule on glia, AMOGsodium/potassium-dependent ATPase beta-2 subunit, ATPase, Na+/K+ transporting, beta 2 polypeptide, Na, K-ATPase beta-2 polypeptide, Sodium/potassium-dependent ATPase subunit beta-2, sodium/potassium-transporting ATPase beta-2 chain, sodium/potassium-transporting ATPase subunit beta-2 | |
Endocrinology, Signal Transduction | |
ATPase Na+/K+ beta 2 |
Product Title
Novus Biologicals™ Recombinant Human ATPase Na+/K+ beta 2 His Protein