missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Carbonic Anhydrase VII Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 3177-CA-050

283.36 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18187480

  • 360.00€ / 50µg
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Recombinant Human Carbonic Anhydrase VII Protein is derived from E. coli. The Recombinant Human Carbonic Anhydrase VII Protein has been validated for the following applications: Enzyme Activity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in Tris and NaCl.
31 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Recombinant
766
Carbonic Anhydrase VII/CA7
E. coli-derived human Carbonic Anhydrase VII/CA7 protein Thr2-Ala264,with a C-terminal 10-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers