missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Carbonic Anhydrase VIII Protein
Brand: R&D Systems™ 2187-CA-050
234.12 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human Carbonic Anhydrase VIII Protein is derived from E. coli.Specifications
Supplied as a 0.2μm filtered solution in Tris and NaCl. | |
34 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. | |
Recombinant |
767 | |
Carbonic Anhydrase VIII/CA8 | |
E. coli-derived human Carbonic Anhydrase VIII/CA8 protein Ala2-Gln290,with a C-terminal 10-His tag | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |