missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Carbonic Anhydrase XII Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 2190-CA-010

234.12 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18167710

  • 297.00€ / 10µg
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Recombinant Human Carbonic Anhydrase XII Protein is derived from NS0. The Recombinant Human Carbonic Anhydrase XII Protein has been validated for the following applications: Enzyme Activity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in Tris and NaCl.
32 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Recombinant
771
Carbonic Anhydrase XII/CA12
Mouse myeloma cell line,NS0-derived human Carbonic Anhydrase XII/CA12 protein Ala25-Gln291,with a C-terminal 10-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers