missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human CSK His Protein
Highly purified. Generating reliable and reproducible results.
394.00€ - 1070.00€
Specifications
Concentration | 0.5mg/mL |
---|---|
For Use With (Application) | SDS-PAGE |
Formulation | Phosphate buffered saline (pH 7.4) containing 20% glycerol, 1mM DTT |
Gene ID (Entrez) | 1445 |
Molecular Weight (g/mol) | TMW: 53.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18256900
|
Novus Biologicals™
NBP2-51853-0.05MG |
0.05 mg |
394.00€
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18294090
|
Novus Biologicals™
NBP2-51853-0.25MG |
0.25 mg |
1070.00€
0.25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-450 of Human CSK The Recombinant Human CSK Protein is derived from E. coli. The Recombinant Human CSK Protein has been validated for the following applications: SDS-Page.Specifications
0.5mg/mL | |
Phosphate buffered saline (pH 7.4) containing 20% glycerol, 1mM DTT | |
TMW: 53.1kDa | |
C-Src kinase, c-src tyrosine kinase, EC 2.7.10, EC 2.7.10.2, MGC117393, Protein-tyrosine kinase CYL, tyrosine-protein kinase CSK | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain |
SDS-PAGE | |
1445 | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
CSK | |
Recombinant |
For research use only and is not approved for use in humans or in clinical diagnosis
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Human CSK His Protein