missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human FAP Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 3715-SE-010
This item is not returnable.
View return policy
Description
Measured by its ability to convert the substrate benzyloxycarbonyl-Gly-Pro-7-amido-4-methylcoumarin (Z-GP-AMC) to Z-Gly-Pro and 7-amino-4-methylcoumarin (AMC). The specific activity is >1800 pmol/min/μg, as measured under the described conditions.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Q12884 | |
Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. | |
M.W. (Observed): 85 kDa, reducing conditions; M.W. (theoretical): 86 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. | |
FAP | |
Recombinant | |
Fibroblast Activation Protein alpha/FAP |
Enzyme Activity | |
2191 | |
Fibroblast Activation Protein alpha/FAP | |
Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Fibroblast Activation Protein alpha/FAP protein Leu26-Asp760, with an N-terminal 6-His tag | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Human FAP Protein > 10ug; Carrier Free
Spot an opportunity for improvement?Share a Content Correction