missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human GFAP Protein
1050.00€ - 1590.00€
Specifications
Formulation | Lyophilized from 16 mM NaHCO3, containing 0.05% CHAPS and 0.05% Tween 20 |
---|---|
Gene ID (Entrez) | 2670 |
Molecular Weight (g/mol) | 37.9 kDa |
Storage Requirements | Store at -70°C. Avoid freeze-thaw cycles. |
Gene Alias | ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18392246
|
Novus Biologicals™
NBP3-11964-0.5MG |
0.5 mg |
1050.00€
0.50mg |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
18328842
|
Novus Biologicals™
NBP3-11964-1MG |
1 mg |
1590.00€
1mg |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
Specifications
Lyophilized from 16 mM NaHCO3, containing 0.05% CHAPS and 0.05% Tween 20 | |
37.9 kDa | |
ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein | |
It is recommended to reconstitute this product with deionized water to its initial concentration. | |
GFAP |
2670 | |
Store at -70°C. Avoid freeze-thaw cycles. | |
GFAP | |
>90%, by SDS-PAGE |