missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Glycoprotein V/CD42d Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 4249-GP-050

325.70 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18357727

  • 426.00€ / 50µg
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Recombinant Human Glycoprotein V/CD42d Protein is derived from NS0. The Recombinant Human Glycoprotein V/CD42d Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
56 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
2814
Glycoprotein V/CD42d
Mouse myeloma cell line,NS0-derived human Glycoprotein V/CD42d protein Gln17-Gly523,with a C-termninal 6-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers