missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Integrin alpha 9 beta 1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 5438-A9-050
373.21 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human Integrin alpha 9 beta 1 Protein is derived from CHO. The Recombinant Human Integrin alpha 9 beta 1 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
114 kDa ( alpha9) & 86.3 kDa (beta1) | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
3680 | |
Integrin alpha 9 beta 1 | |
Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha 9 beta 1 protein, (N-terminus) Human Integrin alpha 9 (Tyr30-Tyr977) Accession &Num; Q13797 Acidic Tail HHHHHH Human Integrin beta 1 (Gln21-Asp728) Accession &Num; P05556 Basic Tail (C-terminus) | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |