missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MOG Protein

A cDNA sequence encoding the MOG was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12718-1mg

 View more versions of this product

Product Code. 15901329

  • 2330.00€ / 1mg
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

MOG Protein
Research Use Only
Recombinant Protein
E. coli
MGQFRVIGPRHPIRALVGDEVELPCRISPGKNATGMEVGWYRPPFSRVVHLYRNGKDQDGDQAPEYRGRTELLKDAIGEGKVTLRIRNVRFSDEGGFTCFFRDHSYQEEAAMELKVEDPFYWVSPGHHHHHH
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The Myelin Oligodendrocyte Glycoprotein 0.5 mg/ml solution was lyophilized from 20mM sodium acetate buffer pH 4 and 0.3M sodium chloride.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers