missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Polycystic kidney disease protein 1-like 2 GST (N-Term) Protein
Recombinant Protein
630.00€ - 801.00€
Specifications
For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
---|---|
Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
Gene ID (Entrez) | 114780 |
Molecular Weight (g/mol) | MolecularWeight-theroretical: 59.4 kDa |
Storage Requirements | Store at -80°C. Avoid freeze/thaw cycles. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18646760
|
Novus Biologicals™
H00114780-P01-10ug |
10 μg |
630.00€
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18699120
|
Novus Biologicals™
H00114780-P01-25ug |
25 μg |
801.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.Specifications
Western Blot, ELISA, Protein Array, Immunoaffinity Purification | |
114780 | |
Store at -80°C. Avoid freeze/thaw cycles. | |
PKD1L2 | |
>80% by SDS-PAGE and Coomassie blue staining |
50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
MolecularWeight-theroretical: 59.4 kDa | |
DKFZp686J19100, FLJ45333, KIAA1879, PC1L2, PC1-like 2 protein, polycystic kidney disease 1-like 2, polycystic kidney disease protein 1-like 2, polycystin-1L2 | |
Neuroscience, Signal Transduction | |
Polycystic kidney disease protein 1-like 2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Human Polycystic kidney disease protein 1-like 2 GST (N-Term) Protein