missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Prosurfactant Protein C GST (N-Term) Protein

Recombinant Protein

630.00€ - 801.00€

Specifications

For Use With (Application) Western Blot, ELISA, PAGE, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 6440
Molecular Weight (g/mol) 47.3 kDa
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18392609
View Documents
Novus Biologicals™
H00006440-P01-10ug
10 μg
630.00€
10µg
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
18347429
View Documents
Novus Biologicals™
H00006440-P01-25ug
25 μg
801.00€
25µg
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
Description

Description

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications

Specifications

Western Blot, ELISA, PAGE, Protein Array, Immunoaffinity Purification
6440
Store at -80°C. Avoid freeze-thaw cycles.
SFTPC
Prosurfactant Protein C
50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
47.3 kDa
PSP-C, pulmonary surfactant-associated protein C, Pulmonary surfactant-associated proteolipid SPL(Val), SFTP2, SFTP2pulmonary surfactant apoprotein-2 SP-C, SMDP2surfactant, pulmonary-associated protein C, SP-CSP5, surfactant protein C
>80% by SDS-PAGE and Coomassie blue staining
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.