Learn More
R&D Systems™ Recombinant Human Relaxin R1 Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 7996-RR-050
Product Code. 18187130
  • 402.00€ / 50µg
Estimated Shipment: 15-01-2025
Description
The Recombinant Human Relaxin R1 Fc Chimera Protein is derived from CHO. The Recombinant Human Relaxin R1 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
69.8 kDa (monomer)
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE with silver staining.
59350
Relaxin R1 Fc Chimera
Chinese Hamster Ovary cell line,CHO-derived human Relaxin R1 protein, (N-terminus) Human Relaxin R1 (Gln23-Ser398) Accession &Num; Q9HBX9 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Disulfide-linked homodimer
Recombinant
Product Suggestions