Learn More
R&D Systems™ Recombinant Human SMOC-2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 5140-SM-050
443.18 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18147830
  • 571.00€ / 50µg
Estimated Shipment: 05-12-2024
Description
The Recombinant Human SMOC-2 Protein is derived from CHO. The Recombinant Human SMOC-2 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
48.3 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
64094
SMOC-2
Chinese Hamster Ovary cell line,CHO-derived human SMOC-2 protein Gln22-Gly446,with a C-terminal 6-His tag
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.