missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Recombinant Human Synaptophysin GST (N-Term) Protein
550.00€ - 739.00€
Specifications
For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification, Block/Neutralize |
---|---|
Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
Gene ID (Entrez) | 6855 |
Molecular Weight (g/mol) | M.W. (Theoretical): 60.17 kDa |
Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18324549
|
Novus Biologicals
H00006855-P01-25ug |
25 μg |
739.00€
25µg |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
18355339
|
Novus Biologicals
H00006855-P01-10ug |
10 μg |
550.00€
10µg |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.Specifications
Western Blot, ELISA, Protein Array, Immunoaffinity Purification, Block/Neutralize | |
6855 | |
Store at -80C. Avoid freeze-thaw cycles. | |
SYP | |
Membrane Vesicle Markers, Stem Cell Markers | |
Synaptophysin |
50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
M.W. (Theoretical): 60.17 kDa | |
Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin | |
Use in blocking/ neutralizing and IHC reported in scientific literature (PMID 24786752). Use in ELISA reported in scientific literature (PMID 23371365). | |
>80% by SDS-PAGE and Coomassie blue staining |