missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Tau 441 Monomer Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Western Blot, In vitro assay, In vivo assay, SDS-Page
623.00€ - 1745.00€
Specifications
For Use With (Application) | Western Blot, In vitro Assay, In vivo Assay, SDS-PAGE |
---|---|
Gene ID (Entrez) | 4137 |
Molecular Weight (g/mol) | 45.8 kDa |
Source | E. coli |
Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18022216
|
Novus Biologicals™
NBP2-76795-100UG |
100 μg |
623.00€
1 set |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
18324400
|
Novus Biologicals™
NBP2-76795-200UG |
200 μg |
859.00€
1 set |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
18361261
|
Novus Biologicals™
NBP2-76795-500UG |
500 μg |
1745.00€
1 set |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
Description
An un-tagged full length Human Recombinant Tau 441 protein Monomer, Accession #: P10636The Recombinant Human Tau 441 Monomer Protein has been validated for the following applications: Western Blot, In vitro assay, In vivo assay, SDS-Page.
Specifications
Western Blot, In vitro Assay, In vivo Assay, SDS-PAGE | |
45.8 kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
MAPT | |
Unconjugated | |
Recombinant | |
0.2 Micrometer Filtered |
4137 | |
E. coli | |
RUO | |
Tau 441 | |
Human | |
10 mM HEPES, 100 mM NaCl pH 7.4 |