missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Tau 441 Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Western Blot, In vitro assay, In vivo assay, SDS-Page

590.00€ - 1745.00€

Specifications

For Use With (Application) Western Blot, In vitro Assay, In vivo Assay, SDS-PAGE
Gene ID (Entrez) 4137
Molecular Weight (g/mol) 45.8 kDa
Source E. coli
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18365430
View Documents
Novus Biologicals™
NBP2-76794-100UG
100 μg
590.00€
1 set
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
18387060
View Documents
Novus Biologicals™
NBP2-76794-200UG
200 μg
834.00€
1 set
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
18293179
View Documents
Novus Biologicals™
NBP2-76794-500UG
500 μg
1745.00€
1 set
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
Description

Description

An un-tagged full length Human Recombinant Tau 441 protein Pre-Formed Fibrils, Accession #: P10636

The Recombinant Human Tau 441 Protein has been validated for the following applications: Western Blot, In vitro assay, In vivo assay, SDS-Page.
Specifications

Specifications

Western Blot, In vitro Assay, In vivo Assay, SDS-PAGE
45.8 kDa
Store at -80°C. Avoid freeze-thaw cycles.
MAPT
Unconjugated
Recombinant
0.2 Micrometer Filtered
4137
E. coli
RUO
Tau 441
Human
10 mM HEPES, 100 mM NaCl pH 7.4
Videos
SDS
Documents

Documents

Product Certifications