missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TLR4/MD-2 Complex Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 3146-TM-050/CF
This item is not returnable.
View return policy
Description
The Recombinant Human TLR4/MD-2 Complex Protein is derived from NS0. The Recombinant Human TLR4/MD-2 Complex Protein has been validated for the following applications: Bioactivity.Specifications
O00206.2 (TLR4) & Q9Y6Y9.1 (MD-2) | |
7099 | |
TLR4/MD-2 Complex | |
Mouse myeloma cell line, NS0-derived human TLR4/MD2 Complex protein Human TLR4 (Glu24-Lys631) Accession # O00206.2(N-terminus) S 10-His tag(C-terminus) Human MD-2 (Glu17-Asn160) Accession # Q9Y6Y9.1 (N-terminus) IEGRGGGSGGGSGGGS 10-His tag (C-terminus ) | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Lyophilized from a 0.2μm filtered solution in PBS. | |
70.6 kDa (TLR4), 19.2 kDa (MD-2) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction