Learn More
R&D Systems™ Recombinant Mouse Carbonic Anhydrase XIV Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 2504-CA-010
234.12 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18107210
  • 297.00€ / 10µg
Estimated Shipment: 09-12-2024
Description
The Recombinant Mouse Carbonic Anhydrase XIV Protein is derived from NS0. The Recombinant Mouse Carbonic Anhydrase XIV Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in Tris and NaCl.
32 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Recombinant
23831
Carbonic Anhydrase XIV/CA14
Mouse myeloma cell line,NS0-derived mouse Carbonic Anhydrase XIV/CA14 protein Ala16-Met290,with a C-terminal 10-His tag Accession &Num; Q9WVT6
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.