missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Cathepsin H Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ 1013-CY-010

346.42 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18137690

  • 440.00€ / 10µg
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Recombinant Mouse Cathepsin H Protein is derived from NS0. The Recombinant Mouse Cathepsin H Protein has been validated for the following applications: Enzyme Activity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in MES and NaCl.
36 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
13036
Cathepsin H
Mouse myeloma cell line,NS0-derived mouse Cathepsin H protein Glu22-Val333,with a C-terminal 10-His tag
Pro form
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers