missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IFNAR2 / IFNABR Protein
A DNA sequence encoding the Mus musculus (Mouse) IFNAR2 / IFNABR, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9199-ye-500ug
This item is not returnable.
View return policy
Specifications
NP_001103968.1 | |
IFNAR2 / IFNABR Protein | |
Research Use Only | |
Ifnar2 | |
Recombinant Protein | |
Yeast | |
SLETITPSAF DGYPDEPCTI NITIRNSRLI LSWELENKSG PPANYTLWYT VMSKDENLTK VKNCSDTTKS SCDVTDKWLE GMESYVVAIV IVHRGDLTVC RCSDYIVPAN APLEPPEFEI VGFTDHINVT MEFPPVTSKI IQEKMKTTPF VIKEQIGDSV RKKHEPKVNN VTGNFTFVLR DLLPKTNYCV SLYFDDDPAI KSPLKCIVLQ PGQESGLSES A | |
Greater than 90% as determined by SDS-PAGE. |
15976 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |