missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IL-19 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

526.00€

Specifications

Gene ID (Entrez) 329244
Molecular Weight (g/mol) 17.7 kDa
Name IL-19
Quantity 25 μg
Source E. coli-derived mouse IL-19 protein Leu25-Ala176,with an N-terminal Met
View More Specs

Products 2
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
18348967
View Documents
R&D Systems™
2915-IL-025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
526.00€
25µg
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
18186170
View Documents
R&D Systems™
2915-IL-025/CF
Lyophilized from a 0.2μm filtered solution in PBS.
526.00€
25µg
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
Description

Description

Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with human IL-20 R alpha and human IL-20 R beta . The ED50 for this effect is 20.0-200 pg/mL.
Specifications

Specifications

329244
IL-19
E. coli-derived mouse IL-19 protein Leu25-Ala176,with an N-terminal Met
IL-10 C, IL19, IL-19, interleukin 19, MDA1, melanoma differentiation associated protein-like protein, NG0.1 Melanoma differentiation-associated protein-like protein, ZMDA1interleukin-19
Recombinant
17.7 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.