missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse LRP-6 N-Terminal Fragment Fc Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 9950-LR-050
293.73 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Mouse LRP-6 N-Terminal Fragment Fc Protein is derived from CHO. The Recombinant Mouse LRP-6 N-Terminal Fragment Fc Protein has been validated for the following applications: Bioactivity.Specifications
NP_032540 | |
Lyophilized from a 0.2 μm filtered solution in PBS. | |
96 kDa | |
Chinese Hamster Ovary cell line,CHO-derived mouse LRP-6 protein, (N-terminus) Mouse LRP-6 (Ala20-Ile628) Accession &Num; NP_032540 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus) | |
RUO | |
Unconjugated | |
Mouse | |
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Bioactivity | |
4040 | |
50 μg | |
12 months from date of receipt, ≤ -20°C as supplied; 1 month, 2 to 8°C under sterile conditions after reconstitution; 3 months, ≤ -20°C under sterile conditions after reconstitution. | |
<0.10 EU per 1 μg of the protein by the LAL method. | |
LRP-6 | |
Reconstitute at 500 μg/mL in PBS. |