missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse MARCO Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand: R&D Systems™ 2956-MA-050
This item is not returnable.
View return policy
Description
The Recombinant Mouse MARCO Protein is derived from NS0. The Recombinant Mouse MARCO Protein has been validated for the following applications: Binding Activity.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
46.3 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
>90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
17167 | |
MARCO | |
Mouse myeloma cell line,NS0-derived mouse MARCO protein Gln70-Ser518,with an N-terminal 9-His tag | |
<0.10 EU per 1μg of the protein by the LAL method. | |
Recombinant |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Mouse MARCO Protein > 50ug; Carrier Free
Spot an opportunity for improvement?Share a Content Correction