missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse p53 / TP53 Protein
A DNA sequence encoding the Mus musculus (Mouse) p53 / TP53, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_001120705.1 |
---|---|
Gene ID (Entrez) | 22059 |
Name | p53 / TP53 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16006090
|
enQuireBio™
QP6823-ec-10ug |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16046090
|
enQuireBio™
QP6823-ec-50ug |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16096080
|
enQuireBio™
QP6823-ec-100ug |
100 μg |
770.00€
100µg |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
16026090
|
enQuireBio™
QP6823-ec-200ug |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16036090
|
enQuireBio™
QP6823-ec-500ug |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16016090
|
enQuireBio™
QP6823-ec-1mg |
1 mg |
3405.00€
1mg |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
Specifications
NP_001120705.1 | |
p53 / TP53 Protein | |
Not determined. | |
Full Length | |
Mouse | |
Tris-based buffer, 50% glycerol |
22059 | |
Research Use Only | |
TP53 | |
Recombinant Protein | |
MEESQSDISL ELPLSQETFS GLWKLLPPED ILPSPHCMDD LLLPQDVEEF FEGPSEALRV SGAPAAQDPV TETPGPVAPA PATPWPLSSF VPSQKTYQGN YGFHLGFLQS GTAKSVMCTY SPPLNKLFCQ LAKTCPVQLW VSATPPAGSR VRAMAIYKKS QHMTEVVRRC PHHERCSDGD GLAPPQHLIR VEGNLYPEYL EDRQTFRHSV VVPYEPPEAG SEYTTIHYKY MCNSSCMGGM NRRPILTIIT LEDSSGNLLG RDSFEVRVCA CPGRDRRTEE ENFRKKEVLC PELPPGSAKR ALPTCTSASP PQKKKPLDGE YFTLKIRGRK RFEMFRELNE ALELKDAHAT EESGDSRAHS SYLKTKKGQS TSRHKKTMVK KVGPDSD |