missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Plasma Kallikrein/KLKB1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 2498-SE-010
This item is not returnable.
View return policy
Description
Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (Catalog # ES002 ). The specific activity is >300 pmol/min/μg, as measured under the described conditions.Specifications
NP_032481 | |
Lyophilized from a 0.2 μm filtered solution in MES and NaCl. | |
M.W. (Observed): 80 kDa, reducing conditions; M.W. (theoretical): 70 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
Reconstitute at 500 μg/mL in sterile 25 mM MES and 150 mM NaCl, pH 5.5. | |
Plasma Kallikrein/KLKB1 |
Enzyme Activity | |
3818 | |
Plasma Kallikrein/KLKB1 | |
Mouse myeloma cell line, NS0-derived mouse Plasma Kallikrein/KLKB1 protein Gly20-Ala638, with a C-terminal 6-His tag | |
Pro form | |
Klkb1 | |
Recombinant | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction