missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Plexin B2 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 6836-PB-050

Product Code. 18300118

  • 483.00€ / 50µg
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Recombinant Mouse Plexin B2 Protein is derived from CHO. The Recombinant Mouse Plexin B2 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
112 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
140570
Plexin B2
Chinese Hamster Ovary cell line,CHO-derived mouse Plexin B2 protein, (N-terminus) Mouse Plexin B2 (Leu20-Trp1029) Accession &Num; NP_001152993 HPGGGSGGGSGGGS 6-His tag (C-terminus)
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.