missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Wnt-9a Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
607.00€
Specifications
Gene ID (Entrez) | 216795 |
---|---|
Molecular Weight (g/mol) | 37 kDa |
Name | Wnt-9a |
Quantity | 25 μg |
Source | Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-9a protein Tyr30-Gly365 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18716977
|
R&D Systems™
8148-WN-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. |
607.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18756987
|
R&D Systems™
8148-WN-025 |
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. |
607.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse Wnt-9a Protein is derived from CHO. The Recombinant Mouse Wnt-9a Protein has been validated for the following applications: Bioactivity.Specifications
216795 | |
Wnt-9a | |
Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-9a protein Tyr30-Gly365 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
37 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Mouse Wnt-9a Protein