missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Zinc finger protein 346 Protein
A DNA sequence encoding the Mus musculus (Mouse) Zinc finger protein 346, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_036147.1 |
---|---|
Gene ID (Entrez) | 26919 |
Name | Zinc finger protein 346 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16006690
|
enQuireBio™
QP6908-ec-10ug |
10 μg | N/A | ||||||
16046690
|
enQuireBio™
QP6908-ec-50ug |
50 μg | N/A | ||||||
16096680
|
enQuireBio™
QP6908-ec-100ug |
100 μg |
770.00€
100µg |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
16026690
|
enQuireBio™
QP6908-ec-200ug |
200 μg | N/A | ||||||
16036690
|
enQuireBio™
QP6908-ec-500ug |
500 μg | N/A | ||||||
16016690
|
enQuireBio™
QP6908-ec-1mg |
1 mg |
3405.00€
1mg |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
Specifications
NP_036147.1 | |
Zinc finger protein 346 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
26919 | |
Research Use Only | |
Znf346 | |
Recombinant Protein | |
E. coli | |
MECPAPDATD AADPGEAGPY KGSEEPEGRE PDGVRFDRER ARRLWEAVSG AQPAGREEVE HMIQKNQCLF TSTQCKVCCA MLISESQKLA HYQSKKHANK VKRYLAIHGM ETIKGDVKRL DSDQKSSRSK DKNHCCPICN MTFSSPAVAQ SHYLGKTHAK SLKLKQQSTK GAALQQNREM LDPDKFCSLC HSTFNDPAMA QQHYMGKRHR KQETKLKLMA HYGRLADPAV SDLPAGKGYP CKTCKIVLNS IEQYQAHVSG FKHKNQSPKT LVTLGSQTPV QTQPTPKDSS TVQD | |
Greater than 90% as determined by SDS-PAGE. |