missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mycoplasma Putative proline iminopeptidase Protein
A DNA sequence encoding the Mycoplasma pneumoniae (strain ATCC 29342 / M129) Putative proline iminopeptidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6964-200ug
This item is not returnable.
View return policy
Specifications
NP_109710.1 | |
Mycoplasma Putative proline iminopeptidase Protein | |
Research Use Only | |
Pip | |
Recombinant Protein | |
E. coli | |
MNTSPKQSGY LKVGNGHEVY FWTAGNPQGK SALYVHGGPG SGTDAGCLKY FDLDTTYVIL LDQRGCGQSK AVNPLLHNTT QDLVGDLEAL RQHLKLERWT LFGGSWGSTL ALVYAITHPQ VVEQVFLRAL FLGREQDWAE MLLGLGKLFY PYEHQTLLKA IPQACRTDFT KFTNYFYEVL QGNDSALKTQ LANAWVKWEN TLLSPISYVK DEKAEDANFT FKLALLECHY AKHHSFLKPN FILENVAVLK DKPVHLIHGR FDLVCPLSQA LELKRALPTL NLYVTNNAGH SGSDPNNLTT IKHLLKTQL | |
Greater than 80% as determined by SDS-PAGE. |
876978 | |
200 μg | |
Not determined. | |
Full Length | |
Mycoplasma | |
His-SUMO | |
Tris-based buffer, 50% glycerol |