Learn More
Thermo Scientific™ Remel™ ADC Enrichment
Remel ADC Enrichment is used in Middlebrook media.
Brand: Thermo Scientific™ R450592
201.77 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Remel ADC Enrichment is used in Middlebrook media.
Specifications
Acetate Differential Agar (Sodium Acetate Agar) | |
D - slant | |
Enrichment | |
20 mL |