missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ ADC Enrichment
Remel ADC Enrichment is used in Middlebrook media.
231.00€ - 259.00€
Specifications
Description | Acetate Differential Agar (Sodium Acetate Agar) |
---|---|
Format | D - slant |
Product Type | Enrichment |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15482913
|
Thermo Scientific™
R450591 |
10 mL | N/A | ||||||
11933262
|
Thermo Scientific™
R450592 |
20 mL |
231.00€
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Remel ADC Enrichment is used in Middlebrook media.
Specifications
Acetate Differential Agar (Sodium Acetate Agar) | |
D - slant | |
Enrichment |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Remel™ ADC Enrichment