Learn More
Thermo Scientific™ Remel™ Contact Plate Sterile Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80 (pH 5.6) (Irradiated)
Thermo Scientific™ Remel Contact Plate Sterile Sabouraud Dextrose Agar with Lecithin, Polysorbate 80 (pH 5.6, Irradiated) is used for cultivation of yeast and fungi.
Brand: Thermo Scientific™ R111805
132.10 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Remel Contact Plate Sterile Sabouraud Dextrose Agar with Lecithin, Polysorbate 80 (pH 5.6) (Irradiated) is used for the cultivation of yeast and fungi.
- Contact Plates Sterile are double bagged plus a 3rd bag is enclosed for customer use (Double + Bag)
- These products are manufactured in a clean room and validated sterile by gamma radiation
- The Contact Plates Sterile listed are made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
Specifications
Contact Plates; Sabouraud Dextrose Agar w/Lecithin; Gamma irradiated | |
Contact - Double Bag | |
Agar | |
10/Pk. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Not intended for IVD use.