Learn More
Thermo Scientific™ Remel™ Contact Plate Sterile Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80 (pH 5.6) (Irradiated)
Thermo Scientific™ Remel Contact Plate Sterile Sabouraud Dextrose Agar with Lecithin, Polysorbate 80 (pH 5.6, Irradiated) is used for cultivation of yeast and fungi.
133.00€
Specifications
Description | Contact Plates; Sabouraud Dextrose Agar w/Lecithin; Gamma irradiated |
---|---|
Format | Contact - Double Bag |
Product Type | Agar |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
17702648
|
Thermo Scientific™
R111805 |
10/Pk. |
133.00€
Pack of 10 |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
Description
Thermo Scientific™ Remel Contact Plate Sterile Sabouraud Dextrose Agar with Lecithin, Polysorbate 80 (pH 5.6) (Irradiated) is used for the cultivation of yeast and fungi.
- Contact Plates Sterile are double bagged plus a 3rd bag is enclosed for customer use (Double + Bag)
- These products are manufactured in a clean room and validated sterile by gamma radiation
- The Contact Plates Sterile listed are made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
Specifications
Contact Plates; Sabouraud Dextrose Agar w/Lecithin; Gamma irradiated | |
Contact - Double Bag | |
Agar |
Not intended for IVD use.
Product Title
Thermo Scientific™ Remel™ Contact Plate Sterile Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80 (pH 5.6) (Irradiated)