Learn More
Macherey-Nagel™ Replacement Batteries
Brand: Macherey-Nagel™ 735500
This item is not returnable.
View return policy
Specifications
Replacement Battery | |
CE Certificate | |
6.4 V |
Lithium-Ion | |
1 Ea. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Product Title
Macherey-Nagel™ Replacement Batteries > Voltage: 6.4V