Learn More
Epredia™ Richard-Allan Scientific™ Amyloid Stain, Sodium Hydroxide Solution with dropper
Demonstrate amyloid with Epredia™ Richard-Allan Scientific™ Amyloid Stain.
Brand:  Epredia™ 88006
 View more versions of this product
Product Code. 12503637
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • Alkaline Congo Red solution will stain amyloid orange-to-red and display apple-green birefringence when examined with polarizing microscope
Specifications
Specifications
Hematoxylin
500mL