Learn More
Epredia™ Richard-Allan Scientific™ Amyloid Stain, Sodium Hydroxide Solution with dropper
Demonstrate amyloid with Epredia™ Richard-Allan Scientific™ Amyloid Stain.
68.49€
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
12503637
|
Epredia™
88006 |
Hematoxylin | N/A | ||||||
Description
- Alkaline Congo Red solution will stain amyloid orange-to-red and display apple-green birefringence when examined with polarizing microscope