man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Facility Safety and Maintenance
Signs and Tags
Facilities Maintenance Signs
Brady™ Rigid Polypropylene Fire Safety Door Signs
Print
Brady™ Rigid Polypropylene Fire Safety Door Signs
14.35€ - 17.58€
See promotional offers below
Products
2
Documents
Product Certifications
Products
2
Product Code
Brand
Dimensions (L x W)
Price
Quantity & Availability
Product Code
Brand
Dimensions (L x W)
Price
Quantity & Availability
11790274
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Brady™
227976
297L x 105 mmH
14.35€
Each
View alternative products
Save Up to
»
Estimated Shipment: 28-10-2024
Log in
to see stock.
Add to basket
11700284
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Brady™
227977
450L x 150 mmH
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
View alternative products
Save Up to
»
Documents
Product Certifications
Back to Top