Learn More
Tocris Bioscience™ RIPK2 inhibitor 1
Potent RIPK2 inhibitor
Brand: Tocris Bioscience™ 6603/50
615.10 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
RIPK2 inhibitor 1 is a potent receptor-interacting protein kinase 2 (RIPK2) inhibitor (IC50 of 5-10 nM). It inhibits NF-κB activity and decreases cancer cell proliferation by 70%. In vivo, RIPK2 inhibitor 1 inhibits intestinal and lung inflammation with low toxicity in rodent models.Specifications
3-(Benzoylamino)-4-methyl-N-[3-(1-methyl-1H-imidazol-2-yl)phenyl]benzamide | |
50 mg | |
C25H22N4O2 |
1290490-78-6 | |
RIP Kinase Inhibitors |