Learn More
STEMCELL Technologies RoboSep™ Human Plasmacytoid DC Isolation Kit
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
Brand:  STEMCELL Technologies 17977RF
Encompass
Product Code. 17157841
  • 2067.00€ / 1 set
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ Human Plasmacytoid DC Isolation Kit is designed to isolate plasmacytoid dendritic cells (pDCs) from human peripheral blood mononuclear cells (PBMCs) or leukapheresis by immunomagnetic negative selection
  • The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
  • The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
  • This product replaces the EasySep™ Human Plasmacytoid DC Enrichment Kit (Catalog #19062), for even faster cell isolations and is compatible with a greater number of EasySep™ magnets
Specifications
Specifications
Dendritic Cells
Cell Isolation
2 x 109 cells