Learn More
STEMCELL Technologies RoboSep™ Human Plasmacytoid DC Isolation Kit
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
Brand: STEMCELL Technologies 17977RF
This item is not returnable.
View return policy
Description
- The EasySep™ Human Plasmacytoid DC Isolation Kit is designed to isolate plasmacytoid dendritic cells (pDCs) from human peripheral blood mononuclear cells (PBMCs) or leukapheresis by immunomagnetic negative selection
- The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
- The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
- This product replaces the EasySep™ Human Plasmacytoid DC Enrichment Kit (Catalog #19062), for even faster cell isolations and is compatible with a greater number of EasySep™ magnets
Specifications
Dendritic Cells | |
Cell Isolation | |
2 x 109 cells |