Learn More
Ohaus™ Rotor, Angle, 44 x 1.5/2.0 ml, ID
Angle Rotor for use with Frontier 5000 Series Multi Pro Centrifuges
Brand: Ohaus™ 30314835
Requires : Frontier 5000 Series Multi Pro Centrifuge
Description
- Capacity: 44 x 1.5/2.0 ml
- Max g-force: 24,039g
- Max speed: 16,000 rpm
Services
2 years
Specifications
Rotor, Angle, 44x1.5/2.0ml, ID | |
44 x 1.5/2.0 mL | |
16,000 rpm | |
FC5714 | |
NA |
Stainless Steel | |
24,039 x g | |
2.410 kg | |
32/55° |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Product Title
Ohaus™ Rotor, Angle, 44 x 1.5/2.0 ml, ID > Model number R-A44 X 2/16MI