Learn More
ITS Thorsten Betzel™ Semiquantitative Strips
Brand:  ITS Thorsten Betzel™ 480002
 View more versions of this product
Product Code. 11702947
  • 23.25€ / Pack of 50
Only null left
Estimated Shipment: 04-07-2025
Add to basket
Specifications
Free Chlorine/0. 1.0. 2.0. 3.0. 4.0. 5.0. 6.0. 7.0ppm(mg/L) and 0.00. 0.05. 0.10. 0.20. 0.40. 0.60. 0.80. 1.20ppm ( mg/L)
Bottle
50 Strips
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
ITS Thorsten Betzel™ Semiquantitative Strips > Free Chlorine Dosage: 0, 0.05, 0.1, 0.2, 0.4, 0.5, 0.8, 1.0, 2.0, 5.0 ppm