Learn More
ITS Thorsten Betzel™ Semiquantitative Strips
18.95€ - 25.30€
Product Code | Brand | Concentration | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Price | Quantity & Availability | ||||||
11752957
|
ITS Thorsten Betzel™
480009 |
Nitrate-Nitrite/NO3: 0. 0.5. 2. 5. 10. 20. 50ppm (mg/L) |
25.30€
Pack of 50 |
Estimated Shipment: 05-05-2025
Log in to see stock. |
||||||
11712947
|
ITS Thorsten Betzel™
481026 |
Free Chlorine/0-0.05-0.1-0.2-0.4-0.6-0. 8-1.2-1.5-2.0-2.6-4.0-> 6.0ppm |
25.30€
Pack of 50 |
Estimated Shipment: 05-05-2025
Log in to see stock. |
||||||
11702947
|
ITS Thorsten Betzel™
480002 |
Free Chlorine/0. 1.0. 2.0. 3.0. 4.0. 5.0. 6.0. 7.0ppm(mg/L) and 0.00. 0.05. 0.10. 0.20. 0.40. 0.60. 0.80. 1.20ppm ( mg/L) |
23.25€
Pack of 50 |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
11742947
|
ITS Thorsten Betzel™
480010 |
Total Chlorine/ 0-0.05-0.1-0.15-0.2-0.5-0.8-1.0-4.0-10.0ppm |
21.75€
Pack of 50 |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
11752947
|
ITS Thorsten Betzel™
480033 |
Total Chlorine/0-5-10-20-30-40-60-80ppm |
18.95€
Pack of 50 |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
11772957
|
ITS Thorsten Betzel™
481015 |
Hydrogen Peroxide/0.05-0.3-0.5-1.0-2.0-4.0ppm |
25.30€
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||||
11762947
|
ITS Thorsten Betzel™
481027 |
Chloride/0-50-100-250-500ppm |
23.25€
Pack of 50 |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
Specifications
Bottle | |
50 Strips |
Product Title
ITS Thorsten Betzel™ Semiquantitative Strips