Learn More
ITS Thorsten Betzel™ Semiquantitative Strips
Brand:  ITS Thorsten Betzel™
 View more versions of this product
Product Code. 11742947
  • 21.75€ / Pack of 50
Estimated Shipment: 13-06-2025
Explore more special offers
Add to basket
Specifications
Total Chlorine/ 0-0.05-0.1-0.15-0.2-0.5-0.8-1.0-4.0-10.0ppm
Bottle
50 Strips
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
ITS Thorsten Betzel™ Semiquantitative Strips > Total Chlorine Dosage: 0, 0.05, 0.1, 0.15, 0.2, 0.5, 0.8, 1.0, 4.0, 10.0ppm