missing translation for 'onlineSavingsMsg'
Learn More

ITS Thorsten Betzel™ Semiquantitative Strips

Brand:  ITS Thorsten Betzel™ 481026

 View more versions of this product

Product Code. 11712947

  • 25.30€ / Pack of 50
In Stock
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Free Chlorine/0-0.05-0.1-0.2-0.4-0.6-0. 8-1.2-1.5-2.0-2.6-4.0-> 6.0ppm
Bottle
50 Strips
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ITS Thorsten Betzel™ Semiquantitative Strips > Free Chlorine Dosage: 0, 0.1, 0.2, 0.5, 1.0, 2.5, 5.0ppm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.