Learn More
Fisherbrand™ Septum for 13-425 screw cap
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
Brand: Fisherbrand™ 12 02 0143
279.72 EUR valid until 2025-04-30
Use promo code "24163" to get your promotional price.
Description
- 12mm Septa
Specifications
Septa | |
12 mm | |
1000 |
Cream, Red | |
PTFE, Silicone | |
1.5 mm |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
58Hours
9Minutes
27Seconds
35New Offer Save Up to 70% -
Fisherbrand
Fisherbrand Sweet Deals on Top Products
Get up to 60% off select Fisherbrand productsPromo Code: 24163Expires: 2025-04-30
Days
27Hours
9Minutes
27Seconds
35