Learn More
Heidolph Set Glass Sensor
Modular temperature sensor consisting of a standard PT1000 (V4A) and a glass coating when working with aggressive media.
Brand:  Heidolph 509-67970-00
Product Code. 17900390
  • 200.00€ / Each
Estimated Shipment: 23-01-2025
Description
  • Modular temperature sensor consisting of a standard PT1000 (V4A) and a glass coating when working with aggressive media.
  • Compared to other glass sensors, you will only have to change the glass coating when it comes to glass breakage and not the whole temperature sensor.
  • Compatible with: Hei-PLATE Mix 'n' Heat Core+, Expert, Ultimate
Specifications
Specifications
Temperature Probe
2 Years
Plastic