Learn More
Epredia™ Double Cytoslide™ Microscope Slides
Use Epredia™ Shandon™ Double Cytoslides™ with Epredia™ Shandon™ EZ Double Cytofunnel™ Chambers.
Brand: Epredia™ 5991054
84.64 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Specimen is displayed within a circle marked as 1 or 2
- Number circles correspond with the Cytofunnel™ chamber
Ordering Alert:
- Sold in boxes of 100 slides
Specifications
Double Cytoslide | |
Plain | |
Noncoated |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%